Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Bioss
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
SH3GL3 is implicated in endocytosis. It may recruit other proteins to membranes with high curvature.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EEN-B2; Endophilin 3; endophilin A3; endophilin III; Endophilin-3; Endophilin-A3; SH3 domain containing GRB2 like 3; SH3 domain containing GRB2 like endophilin A3; SH3 domain protein 2 C1; SH3 domain protein 2 C2; SH3 domain protein 2C; SH3 domain-containing GRB2-like protein 3; SH3-domain GRB2-like 3; SH3p13
Gene Aliases: CNSA3; EEN-B2; HsT19371; SH3D2C; Sh3d2c1; Sh3d2c2; SH3GL3; SH3P13
UniProt ID: (Human) Q99963, (Mouse) Q62421, (Rat) O35180
Entrez Gene ID: (Human) 6457, (Mouse) 20408, (Rat) 81921
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support