Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: EVLLLHMCSY LDMRALGRLA QVYRWLWHFT NCDLLRRQIA WASLNSGFTR LGTNLMTSVP VKVSQNWIVG CCREGILLKW RCSQMPWMQL EDDALYISQA
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Gene Aliases: FBW3; FBXW3; SHFM3P1
Entrez Gene ID: (Human) 26226
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support