Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MAARPITLGI DLGTTSVKAA LLRAAPDDPS GFAVLASCAR AARAEAAVES AVAGPQGREQ DVSRILQALH ECLAALPRPQ LRSVVGIGVS GQMHGVVFWK TGQGCEWTEG GITPVFEPRA VSHLVTWQDG RCSSEFLASL PQPKSHLSVA TGFGCATIFW LLKYRPEFLK SYDAAGTIHD YVVAMLCGLP RPLMSDQNAA SWGYFNTQSQ SWNVETLRSS GFPVHLLPDI AEPGSVAGRT SHMWFEIPKG TQVGVALGDL QASVYSCMAQ RTDAVLNIST SVQLAASMPS GFQPAQTPDP TAPVAYFPYF NRTYLGVAAS LNGGNVLATF VHMLVQWMAD LGLEVEESTV YSRMIQAAVQ QRDTHLTITP TVLGERHLPD QLASVTRISS SDLSLGHVTR ALCRGIVQNL HSMLPIQQLQ EWGVERVMGS GSALSRNDVL KQEVQRAFPL PMSFGQDVDA AVGAALVMLR RHLNQKES
The protein encoded by this gene has weak homology to several carbohydrate kinases, a class of proteins involved in the phosphorylation of sugars as they enter a cell, inhibiting return across the cell membrane. Sequence variation between this novel gene and known carbohydrate kinases suggests the possibility of a different substrate, cofactor or changes in kinetic properties distinguishing it from other carbohydrate kinases. The gene resides in a region commonly deleted in cystinosis patients, suggesting a role as a modifier for the cystinosis phenotype. The genomic region is also rich in Alu repetitive sequences, frequently involved in chromosomal rearrangements.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Carbohydrate kinase-like protein; Sedoheptulokinase; SHK
Gene Aliases: CARKL; SHK; SHPK
UniProt ID: (Human) Q9UHJ6
Entrez Gene ID: (Human) 23729
Molecular Function: carbohydrate kinase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support