Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: IFRNGIALE ILQNTSYLPV LEGQALRLLC DAPSNPPAHL SWFQGSPALN ATPISNTGIL ELRRVRSAEE GGFTCRAQHP LGFLQIFLNL SVYSLPQLLG PSCSWEAEGL HCRCSFRARP APSLCWRLEE KPLEGNSSQG SFKVNSSSAG PWANSSLILH GGLSSDLKVS CKAWNIYGSQ SGSVLLLQGR SNLGTGVVPA ALG (241-442 aa encoded by BC029896)
SIGLEC5 is a putative adhesion molecule that mediates sialic-acid dependent binding to cells. This protein binds equally to alpha2,3-linked and alpha2,6-linked sialic acid. The sialic acid recognition site may be masked by cis interactions with sialic acids on the same cell surface. This molecule is expressed by monocytic/myeloid lineage cells. SIGLEC5 is found at high levels in peripheral blood leukocytes, spleen, bone marrow and at lower levels in lymph node, lung, appendix, placenta, pancreas and thymus. It is expressed by monocytes and neutrophils but absent from leukemic cell lines representing early stages of myelomonocytic differentiation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD170; CD33 antigen-like 2; OB-binding protein 2; OB-BP2; Obesity-binding protein 2; sialic acid binding Ig-like lectin 5; Sialic acid-binding Ig-like lectin 5; sialic acid-binding immunoglobulin-like lectin 5; Siglec; Siglec-5
Gene Aliases: CD170; CD33L2; OB-BP2; OBBP2; SIGLEC-5; SIGLEC5
UniProt ID: (Human) O15389
Entrez Gene ID: (Human) 8778
Molecular Function:
immunoglobulin superfamily cell adhesion molecule
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support