Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MQGAQEASAS EMLPLLLPLL WAGALAQERR FQLEGPESLT VQEGLCVLVP CRLPTTLPAS YYGYGYWFLE GADVPVATND PDEEVQEETR GRFHLLWDPR RKNCSLSIRD ARRRDNAAYF FRLKSKWMKY GYTSSKLSVR VMALTHRPNI SIPGTLESGH PSNLTCSVPW VCEQGTPPIF SWMSAAPTSL GPRTTQSSVL TITPRPQDHS TNLTCQVTFP GAGVTMERTI QLNVSSFKIL QNTSSLPVLE GQALRLLCDA DGNPPAHLSW FQGFPALNAT PISNTGVLEL PQVGSAEEGD FTCRAQHPLG SLQISLSLFV HWKPEGRAGG VLGAVWGASI TTLVFLCVCF IFRVKTRRKK AAQPVQNTDD VNPVMVSGSR GHQHQFQTGI VSDHPAEAGP ISEDEQELHY AVLHFHKVQP QEPKVTDTEY SEIKIHK
SIGLEC9 (Sialic acid-binding Ig-like lectin 9) is a putative adhesion molecule that mediates sialic-acid dependent binding to cells. It contains two Ig-like C2 type (immunoglobulin-like) domains and one Ig-like V-type (immunoglobulin-like) domain. It preferentially binds to alpha2, 3-or alpha2, 6-linked sialic acid. The sialic acid recognition site may be mased by cis interactions with sialic acids on the same cell surface.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD327; CD33 antigen-like 1; CDw327; OB-BP1; Obesity-binding protein 1; sialic acid binding Ig-like lectin 6; Sialic acid-binding Ig-like lectin 6; Siglec; Siglec-6
Gene Aliases: CD327; CD33L; CD33L1; CD33L2; CDW327; OBBP1; SIGLEC6
UniProt ID: (Human) O43699
Entrez Gene ID: (Human) 946
Molecular Function: immunoglobulin superfamily cell adhesion molecule
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support