Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
In direct ELISAs, approximately 20% cross-reactivity with recombinant mouse (rm) SIGNR3 and rmSIGNR4 is observed, and approximately 10% cross-reactivity with recombinant human DC-SIGNR and rmDC-SIGN is observed.
Reconstitute at 0.2 mg/mL in sterile PBS.
Endoxin level is <0.10 EU per 1 µg of the antibody by the LAL method.
SIGNR1 is a type II transmembrane C-type lectin that was identified in a search for mouse homologues of human DC-SIGN. It is expressed at high levels in splenic marginal zone macrophages and lymph node medullary macrophages, where it functions to uptake dextran polysaccharides, including the capsular polysaccharide of Streptococcus pneumoniae. It has also been demonstrated that SIGNR1 physically associates with TLR4/MD2, and it has been suggested that this association plays a role in recognition of LPS. Furthermore, recently it has been shown that SIGNR1 deficient mice have a defect in catabolism of the complement component C3, and that SIGNR1 binds directly to the complement C1 subcomponent, C1q to assemble a non-conventional C3 convertase.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD209; CD209 antigen-like protein A; CD209 antigen-like protein B; DC SIGNR1; DC-SIGN; DC-SIGN-related protein 1; DC-SIGNR1; Dendritic cell-specific ICAM-3-grabbing non-integrin; MGC130443; OtB7
Gene Aliases: 1810030I22Rik; CD209; Cd209a; Cd209b; CDSIGN; CIRE; DC-SIGN; DC-SIGN1; DC-SIGNR1; Dcsign; mSIGNR1; OtB7; SIGN-R1; SIGNR1; SIGNR5
UniProt ID: (Mouse) Q91ZX1, (Mouse) Q8CJ91
Entrez Gene ID: (Mouse) 170786, (Mouse) 69165
Molecular Function:
membrane traffic protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support