Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MVIMSEFSAD PAGQGQGQQK PLRVGFYDIE RTLGKGNFAV VKLARHRVTK TQVAIKIIDK TRLDSSNLEK IYREVQLMKL LNHPHIIKLY QVMETKDMLY IVTEFAKNGE MFDYLTSNGH LSENEARKKF WQILSAVEYC HDHHIVHRDL KTENLLLDGN MDIKLADFGF GNFYKSGEPL STWCGSPPYA APEVFEGKEY EGPQLDIWSL GVVLYVLVCG SLPFDGPNLP TLRQRVLEGR FRIPFFMSQD CESLIRRMLV VDPARRITIA QIRQHRWMRA EPCLPGPACP AFSAHSYTSN LGDYDEQALG IMQTLGVDRQ RTVESLQNSS YNHFAAIYYL LLERLKEYRN AQCARPGPAR QPRPRSSDLS GLEVPQEGLS TDPFRPALLC PQPQTLVQSV LQAEMDCELQ SSLQWPLFFP VDASCSGVFR PRPVSPSSLL DTAISEEARQ GPGLEEEQDT QESLPSSTGR RHTLAEVSTR LSPLTAPCIV VSPSTTASPA EGTSSDSCLT FSASKSPAGL SGTPATQGLL GACSPVRLAS PFLGSQSATP VLQAQGGLGG AVLLPVSFQE GRRASDTSLT QGLKAFRQQL RKTTRTKGFL GLNKIKGLAR QVCQVPASRA SRGGLSPFHA PAQSPGLHGG AAGSREGWSL LEEVLEQQRL LQLQHHPAAA PGCSQAPQPA PAPFVIAPCD GPGAAPLPST LLTSGLPLLP PPLLQTGASP VASAAQLLDT HLHIGTGPTA LPAVPPPRLA RLAPGCEPLG LLQGDCEMED LMPCSLGTFV LVQ
SIK1 may mediate the physiological effects of LKB1, including its tumor suppressor function. SIK1 is also involved in signaling by various proteins like STRAD, NUAK1, NUAK2, BRSK1, BRSK2, QIK, QSK, SIK, MARK1, MARK2, MARK3, MARK4, and MELK that are related to AMPK. Activation of SIK1 by phosphorylation on thr322 can lead to an increase in the catalytic activity of sodium/potassium ATPase alpha subunit at the plasma membrane. This results in an increase in intracellular sodium in intact mammalian cells. The family has been classified in 8 major groups based on sequence comparison of their tyrosine (PTK) or serine/threonine (STK) kinase catalytic domains. The STE group (homologs of yeast Sterile 7, 11, 20 kinases) consists of 50 kinases related to the mitogen-activated protein kinase (MAPK) cascade families (Ste7/MAP2K, Ste11/MAP3K, and Ste20/MAP4K). MAP kinase cascades, consisting of a MAPK and one or more upstream regulatory kinases (MAPKKs) have been best characterized in the yeast pheromone response pathway. Pheromones bind to Ste cell surface receptors and activate yeast MAPK pathway.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HRT-20; Myocardial SNF1-like kinase; Probable serine/threonine protein kinase SNF1LK (HRT-20) (Myocardial SNF1-like kinase); Salt-inducible kinase 1; salt-inducible protein kinase 1; serine/threonine protein kinase; Serine/threonine-protein kinase SIK1; Serine/threonine-protein kinase SNF1-like kinase 1; Serine/threonine-protein kinase SNF1LK; SIK-1; SNF1-like kinase
Gene Aliases: EIEE30; MSK; SIK; SIK1; SNF1LK
UniProt ID: (Human) P57059, (Mouse) Q60670
Entrez Gene ID: (Human) 150094, (Mouse) 17691
Molecular Function:
non-receptor serine/threonine protein kinase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support