Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: AHAFEAFQST RSGQRRHTLS EVTNQLVVMP GAGKIFSMND SPSLDSVDSE YDMGSVQRDL NFLEDNPSLK DIMLANQPSP RMTSPFISLR PTNPAMQALS SQKREVHNRS
SNF1LK2 (QIK) is a serine/threonine kinase and is a salt-inducible kinase that belongs to the AMP-activated protein kinase family. SNF1LK2 negatively regulates CRE-binding protein (CREB) activity by phosphorylating the CREB-specific coactivator transducer of regulated CREB activity (TORC). SIK2 is thought to be part of a signaling cascade that regulates the expression and activity of the insulin-induced genes PGC-1'alpha and UCP-1 in brown adipocytes, impairment of which has been implicated in obesity and insulin resistance in human and animal models. SIK2 has also been reported as a key regulator for neuronal survival after ischemia, suppressing CREB-mediated gene expression after oxygen-glucose deprivation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Qin-induced kinase; Salt-inducible kinase 2; salt-inducible protein kinase 2; salt-inducible serine/threonine kinase 2; Serine/threonine-protein kinase SIK2; Serine/threonine-protein kinase SNF1-like kinase 2; SIK-2; SNF1-like kinase 2
Gene Aliases: KIAA0781; LOH11CR1I; QIK; SIK-2; SIK2; SNF1LK2
UniProt ID: (Human) Q9H0K1
Entrez Gene ID: (Human) 23235
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support