Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
The recommended shelf life for this product is 1 year from date of receipt.
Plays an essential role in early embryonic development.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: SCL-interrupting locus protein; SCL-interrupting locus protein homolog; TAL-1-interrupting locus protein; Tal1 interrupting locus
Gene Aliases: MCPH7; SIL; STIL
UniProt ID: (Human) Q15468, (Mouse) Q60988
Entrez Gene ID: (Human) 6491, (Mouse) 20460
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support