Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MDLVLKRCLL HLAVIGALLA VGATKVPRNQ DWLGVSRQLR TKAWNRQLYP EWTEAQRLDC WRGGQVSLKV SNDGPTLIGA NASFSIALNF PGSQKVLPDG QVIWVNNTII NGSQVWGGQP VYPQETDDAC IFPDGGPCPS GSWSQKRSFV YVWKTWGQYW QVLGGPVSGL SIGTGRAMLG THTMEVTVYH RRGSRSYVPL AHSSSAFTIT DQVPFSVSVS QLRALDGGNK HFLRNQPLTF ALQLHDPSGY LAEADLSYTW DFGDSSGTLI SRALVVTHTY LEPGPVTAQV VLQAAIPLTS CGSSPVPGTT DGHRPTAEAP NTTAGQVPTT EVVGTTPGQA PTAEPSGTTS VQVPTTEVIS TAPVQMPTAE STGMTPEKVP VSEVMGTTLA EMSTPEATGM TPAEVSIVVL SGTTAAQVTT TEWVETTARE LPIPEPEGPD ASSIMSTESI TGSLGPLLDG TATLRLVKRQ VPLDCVLYRY GSFSVTLDIV QGIESAEILQ AVPSGEGDAF ELTVSCQGGL PKEACMEISS PGCQPPAQRL CQPVLPSPAC QLVLHQILKG GSGTYCLNVS LADTNSLAVV STQLIMPGQE AGLGQVPLIV GILLVLMAVV LASLIYRRRL MKQDFSVPQL PHSSSHWLRL PRIFCSCPIG ENSPLLSGQQ V
Melanoma is a malignant tumor of melanocytes which are found predominantly in skin but also in the bowel and the eye (see uveal melanoma). It is one of the rarer types of skin cancer but causes the majority of skin cancer related deaths.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 95kDa melanocyte-specific secreted glycoprotein; ME20-M; ME20M; Melanocyte lineage specific antigen GP100; melanocyte protein mel 17; Melanocyte protein PMEL; Melanocyte protein Pmel 17; Melanocytes lineage-specific antigen GP100; Melanoma-associated ME20 antigen; melanosomal matrix protein17; P1; P100; Pmel 17; Premelanosome protein; Silver locus protein homolog; silver, mouse, homolog of
Gene Aliases: D12S53E; gp100; ME20; ME20-M; ME20M; P1; P100; PMEL; PMEL17; SI; SIL; SILV
UniProt ID: (Human) P40967
Entrez Gene ID: (Human) 6490
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support