Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MAHAGGGSGG SGAGGPAGRG LSGARWGRSG SAGHEKLPVH VEDALTYLDQ VKIRFGSDPA TYNGFLEIMK EFKSQSIDTP GVIRRVSQLF HEHPDLIVGF NAFLPLGYRI DIPKNGKLNI QSPLTSQENS HNHGDGAEDF KQQVPYKEDK PQVPLESDSV EFNNAISYVN KIKTRFLDHP EIYRSFLEIL HTYQKEQLNT RGRPFRGMSE EEVFTEVANL FRGQEDLLSE FGQFLPEAKR SLFTGNGPCE MHSVQKNEHD KTPEHSRKRS RPSLLRPVSA PAKKKMKLRG TKDLSIAAVG KYGTLQEFSF FDKVRRVLKS QEVYENFLRC IALFNQELVS GSELLQLVSP FLG
Acts as a transcriptional repressor. Interacts with MXI1 to repress MYC responsive genes and antagonize MYC oncogenic activities. Interacts with MAD-MAX heterodimers by binding to MAD. The heterodimer then represses transcription by tethering SIN3B to DNA. Also forms a complex with FOXK1 which represses transcription.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Histone deacetylase complex subunit Sin3b; Paired amphipathic helix protein Sin3b; SIN3 homolog B, transcriptional regulator; SIN3 transcription regulator homolog B; SIN3B; Transcriptional corepressor Sin3b
Gene Aliases: KIAA0700; SIN3B
UniProt ID: (Human) O75182
Entrez Gene ID: (Human) 23309
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support