Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: MSMLPTFGFT QEQVACVCEV LQQGGNIERL GRFLWSLPAC EHLHKNESVL KAKAVVAFHR GNFRELYKIL ESHQFSPHNH AKLQQLWLKA HYIEAEKLRG RPLGAVGKYR VRRKFPLPRS IWDGEETSYC FKEKSRSVLR EWYAHNPYPS PREKRELTEA TGLTTTQVSN WFKNRRQRDR AAEAKERENN ENSNSNSHNP LNGSGKSVLG SSEDEKTPSG TPDHSSSSPA LLLSPPPPGL PSLHSLGHPP GPSAVPVPVP GGGGADPLQH HHGLQDSILN PMSANLVDLG S
This gene is a member of the vertebrate gene family which encode proteins homologous to the Drosophila 'sine oculis' homeobox protein. SIX2 is a transcription factor which, like other members of this gene family, may be involved in limb or eye development.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Homeobox protein SIX2; Sine oculis homeobox homolog 2
Gene Aliases: SIX2
UniProt ID: (Human) Q9NPC8
Entrez Gene ID: (Human) 10736
Molecular Function: homeodomain transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support