Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: DAFGDELFS VFEGDSTTAA GTKKDKEKDK GKWKGPPGSA DKAGKRFDGK LQSESTNNGK NKRDVDFEGT DEPIFGKKPR IEESITEDLS LADLMPRVKV QSVETVEGCT HEVALPAEED YLPLKPRVGK AAKEYPFILD AFQREAIQCV DNNQSVLVSA HTSAGKTVCA EYAIALALRE KQRVIFTSPI KALSNQKYRE MYEEFQDVGL MTGDVTINPT ASCLVMTTEI LRSMLYRGSE VMREVAWVIF DEIHYMRDSE RGVVWEETII LLPDNVHYVF LSATIPNARQ FAEWICHLHK QPCHVI (3-307 aa encoded by B C028604)
SKIV2L2 may be involved in pre-mRNA splicing.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ATP-dependent helicase SKIV2L2; ATP-dependent RNA helicase DOB1; ATP-dependent RNA helicase SKIV2L2; Exosome RNA helicase MTR4; functional spliceosome-associated protein 118; Superkiller viralicidic activity 2-like 2; TRAMP-like complex helicase
Gene Aliases: 2610528A15Rik; DOB1; fSAP118; KIAA0052; mKIAA0052; MTR4; MTREX; RGD1305984; SKIV2L2
UniProt ID: (Human) P42285, (Mouse) Q9CZU3
Entrez Gene ID: (Human) 23517, (Mouse) 72198, (Rat) 365668
Molecular Function: DNA metabolism protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support