Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Immunogen sequence: MPSIKLQSS DGEIFEVDVE IAKQSVTIKT MLEDLGMDDE GDDDPVPLPN VNAAILKKVI QWCTHHKDDP PPPEDDENKE KRTDDIPVWD QEFLKVDQGT LFELILAANY LDIKGLLDVT CKTVANMIKG KTPEEIRKTF NIKNDFTEEE EAQVRKENQW CEEK (1-163 aa encoded by BC009839)
This gene encodes a component of SCF complexes, which are composed of this protein, cullin 1, a ring-box protein, and one member of the F-box family of proteins. This protein binds directly to the F-box motif found in F-box proteins. SCF complexes are involved in the regulated ubiquitination of specific protein substrates, which targets them for degradation by the proteasome. Specific F-box proteins recognize different target protein(s), and many specific SCF substrates have been identified including regulators of cell cycle progression and development. Studies have also characterized the protein as an RNA polymerase II elongation factor. Alternative splicing of this gene results in two transcript variants. A related pseudogene has been identified on chromosome 7.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cyclin A/CDK2-associated p19; cyclin A/CDK2-associated protein p19; Cyclin-A/CDK2-associated protein p19; MGC34403; OCP-2; OCP-II; OCP-II protein; Organ of Corti protein 2; Organ of Corti protein II; p19A; p19skp1; polypeptide 1-like; RNA polymerase II elongation factor-like protein; RNA polymerase II elongation factor-like protein OCP2; S-phase kinase-associated protein 1; S-phase kinase-associated protein 1A; SIII; Transcription elongation factor B polypeptide 1-like
Gene Aliases: 15kDa; 2610043E24Rik; 2610206H23Rik; EMC19; OCP-II; OCP2; p19A; p19Skp1; SKP1; SKP1A; TCEB1L
UniProt ID: (Human) P63208, (Rat) Q6PEC4, (Mouse) Q9WTX5
Entrez Gene ID: (Human) 6500, (Rat) 287280, (Mouse) 21402
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support