Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Alomone Labs, LTD
Promotions
View available promotion(s)
Promo Code: P5877545 Don't miss out: buy 3, only pay for 2 Extended to June 20: take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
FIGURE: 1 / 4
Reconstitution: 25 µL, 50 µL or 0.2 mL double distilled water (DDW), depending on the sample size. The antibody ships as a lyophilized powder at room temperature. Upon arrival, it should be stored at -20C. The reconstituted solution can be stored at 4C for up to 1 week. For longer periods, small aliquots should be stored at -20C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min).
NRAMP1, also known as SLC11A1, is a member of the solute carrier family 11 (proton-coupled divalent metal ion transporters) family and encodes a multi-pass membrane protein. The protein functions as a divalent transition metal (iron and manganese) transporter involved in iron metabolism and host resistance to certain pathogens. Mutations in this gene have been associated with susceptibility to infectious diseases such as leprosy and tuberculosis, and inflammatory diseases such as Crohn disease and rheumatoid arthritis. Alternatively spliced variants that encode different protein isoforms have been described but the full-length nature of only one has been determined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: host resistance locus Bcg/Ity/Lsh; natural resistance associated macrophage protein 1; Natural resistance-associated macrophage protein 1; NRAMP 1; solute carrier family 11 (proton-coupled divalent metal ion transporter), member 1; solute carrier family 11 (proton-coupled divalent metal ion transporters), member 1; solute carrier family 11 (sodium/phosphate symporters), member 1; Solute carrier family 11 member 1
Gene Aliases: Bcg; Itg; Ity; LSH; NRAMP; NRAMP1; SLC11A1
UniProt ID: (Human) P49279, (Mouse) P41251, (Rat) P70553
Entrez Gene ID: (Human) 6556, (Mouse) 18173, (Rat) 316519
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support