Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
Antibody detects endogenous levels of total SLC13A2.
SLC13A2 belongs to the SLC13A transporter (TC 2.A.47) family, NADC subfamily. It is a multi-pass membrane protein. SLC13A2 cotransports of sodium ions and dicarboxylates such as succinate and citrate.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Intestinal sodium/dicarboxylate cotransporter; Na(+)-coupled citrate transporter; Na(+)/dicarboxylate cotransporter 1; NaDC-1; Renal sodium/dicarboxylate cotransporter; sodium-dependent dicarboxylate transporter; sodium/dicarboxylate co-transporter; solute carrier family 13 (sodium-dependent dicarboxylate transporter), member 2; Solute carrier family 13 member 2; solute carrier family 13, member 2
Gene Aliases: mNaDC-1; mucin; NaCT; NaDC-1; NADC1; SDCT1; SLC13A2
UniProt ID: (Human) Q13183, (Rat) P70545, (Mouse) Q9ES88
Entrez Gene ID: (Human) 9058, (Rat) 65202, (Mouse) 20500
Molecular Function:
transporter
secondary carrier transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support