Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: VEAILQQMEA TSAATEAGLE LVDKGKAKEL PGSQVIFEGP TLGQQEDQER KRLCK
S13A5 is a tricarboxylate plasma transporter with a preference for citrate.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Na(+)/citrate cotransporter; Na+-coupled citrate transporter protein; NaCT; Sodium-coupled citrate transporter; Sodium-dependent citrate transporter; sodium-dependent dicarboxylate transporter; solute carrier family 13 (sodium-dependent citrate transporter), member 5; Solute carrier family 13 member 5
Gene Aliases: EIEE25; mIndy; NACT; SLC13A5
UniProt ID: (Human) Q86YT5
Entrez Gene ID: (Human) 284111
Molecular Function: transporter secondary carrier transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support