Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MPPAVGGPVG YTPPDGGWGW AVVIGAFISI GFSYAFPKSI TVFFKEIEGI FHATTSEVSW ISSIMLAVMY GGGPISSILV NKYGSRIVMI VGGCLSGCGL IAASFCNTVQ QLYVCIGVIG GLGLAFNLNP ALTMIGKYFY KRRPLANGLA MAGSPVFLCT LAPLNQVFFG IFGWRGSFLI LGGLLLNCCV AGALMRPIGP KPTKAGKDKS KASLEKAGKS GVKKDLHDAN TDLIGRHPKQ EKRSVFQTIN QFLDLTLFTH RGFLLYLSGN VIMFFGLFAP LVFLSSYGKS QHYSSEKSAF LLSILAFVDM VARPSMGLVA NTKPIRPRIQ YFFAASVVAN GVCHMLAPLS TTYVGFCVYA GFFGFAFGWL SSVLFETLMD LVGPQRFSSA VGLVTIVECC PVLLGPPLLG RLNDMYGDYK YTYWACGVVL IISGIYLFIG MGINYRLLAK EQKANEQKKE SKEEETSIDV AGKPNEVTKA AESPDQKDTE GGPKEEESPV
SLC16A1 is a monocarboxylate transporter (MCT1) that mediates the movement of lactate and pyruvate across cell membranes. Import and export of these substrates by tissues such as erythrocytes, muscle, intestine, and kidney are ascribed largely to the action of a proton-coupled MCT.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ36745; MCT 1; MGC44475; monocarboxylate transporter; Monocarboxylate transporter 1; SLC16A1 protein; solute carrier family 16 (monocarboxylate transporter), member 1; solute carrier family 16 (monocarboxylic acid transporters), member 1; Solute carrier family 16 member 1; solute carrier family 16, member 1 (monocarboxylic acid transporter 1)
Gene Aliases: HHF7; MCT; MCT1; MCT1D; SLC16A1
UniProt ID: (Human) P53985
Entrez Gene ID: (Human) 6566
Molecular Function:
transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support