Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Antibody detects endogenous levels of total MOT11.
Proton-linked monocarboxylate transporter. It catalyzes the transport of pyruvate across the plasma membrane (PubMed:28666119). Probably involved in hepatic lipid metabolism: overexpression results in an increase of triacylglycerol(TAG) levels, small increases in intracellular diacylglycerols and decreases in lysophosphatidylcholine, cholesterol ester and sphingomyelin lipids (PubMed:24390345). [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MCT 11; monocarboxylate transporter; Monocarboxylate transporter 11; monocarboxylic acid transporter 11; monocarboxylic acid transporters; novel RNA polymerase Rpb1 C-terminal repeat domain containing protein; solute carrier family 16 (monocarboxylic acid transporters), member 11; Solute carrier family 16 member 11; solute carrier family 16, member 11; solute carrier family 16, member 11 (monocarboxylic acid transporter 11)
Gene Aliases: AB041591; MCT11; MNCb-2717; SLC16A11
UniProt ID: (Human) Q8NCK7, (Mouse) Q5NC32
Entrez Gene ID: (Human) 162515, (Rat) 287450, (Mouse) 216867
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support