Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The antibody is specific to SLC16A12.
This gene encodes a transmembrane transporter that likely plays a role in monocarboxylic acid transport. A mutation in this gene has been associated with juvenile cataracts with microcornea and renal glucosuria.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Creatine transporter 2; CRT2; MCT 12; monocarboxylate transporter; Monocarboxylate transporter 12; monocarboxylic acid transporter 12; MOT12; solute carrier family 16 (monocarboxylic acid transporters), member 12; Solute carrier family 16 member 12; solute carrier family 16, member 12 (monocarboxylic acid transporter 12)
Gene Aliases: 3230401A21; AW210596; CJMG; CRT2; MCT12; RGD1311468; SLC16A12
UniProt ID: (Human) Q6ZSM3, (Rat) D4A734, (Mouse) Q8BGC3
Entrez Gene ID: (Human) 387700, (Rat) 309525, (Mouse) 240638
Molecular Function: transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support