Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 5
Immunogen sequence: YTSHEDIGYD FEDGPKDKKT LKPHPNIDGG
Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 78%.
Proton-linked monocarboxylate transporter. May catalyze the transport of monocarboxylates across the plasma membrane.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MCT 14; monocarboxylate transporter; Monocarboxylate transporter 14; monocarboxylic acid transporter 14; MOT14; solute carrier family 16 (monocarboxylic acid transporters), member 14; Solute carrier family 16 member 14; solute carrier family 16, member 14 (monocarboxylic acid transporter 14)
Gene Aliases: MCT14; SLC16A14
UniProt ID: (Human) Q7RTX9
Entrez Gene ID: (Human) 151473
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support