Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Predicted to enable monocarboxylic acid transmembrane transporter activity. Involved in urate metabolic process. Predicted to be integral component of plasma membrane.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MCT 9; monocarboxylate transporter; Monocarboxylate transporter 9; monocarboxylic acid transporter 9; solute carrier family 16 (monocarboxylic acid transporters), member 9; Solute carrier family 16 member 9; solute carrier family 16, member 9 (monocarboxylic acid transporter 9)
Gene Aliases: 1200003C15Rik; 4930425B13Rik; AI552636; C10orf36; MCT9; SLC16A9
UniProt ID: (Human) Q7RTY1, (Mouse) Q7TM99
Entrez Gene ID: (Human) 220963, (Mouse) 66859
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support