Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This gene encodes a membrane transporter that exports free sialic acids that have been cleaved off of cell surface lipids and proteins from lysosomes. Mutations in this gene cause sialic acid storage diseases, including infantile sialic acid storage disorder and and Salla disease, an adult form.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AST; FLJ22227; FLJ23268; H(+)/nitrate cotransporter; H(+)/sialic acid cotransporter; hp59; Membrane glycoprotein HP59; proton-coupled sialic acid transporter; sialic acid storage disease; Sialin; sodium/sialic acid cotransporter; solute carrier family 17 (acidic sugar transporter), member 5; Solute carrier family 17 (Anion/sugar transporter), member 5; Solute carrier family 17 member 5; VEAT; Vesicular excitatory amino acid transporter; vesicular H(+)/Aspartate-glutamate cotransporter
Gene Aliases: 4631416G20Rik; 4732491M05; AST; ISSD; NSD; SD; SIALIN; SIASD; SLC17A5; SLD
UniProt ID: (Human) Q9NRA2, (Mouse) Q8BN82, (Rat) Q5Q0U0
Entrez Gene ID: (Human) 26503, (Mouse) 235504, (Rat) 363103
Molecular Function:
secondary carrier transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support