Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Reconstitution: 25 µL, 50 µL or 0.2 mL double distilled water (DDW), depending on the sample size. The antibody ships as a lyophilized powder at room temperature. Upon arrival, it should be stored at -20C. The reconstituted solution can be stored at 4C, protected from the light, for up to 1 week. For longer periods, small aliquots should be stored at -20C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 X g 5 min).
Involved in vesicular storage and exocytosis of ATP. May accumulate ATP and other nucleotides in secretory vesicles such as adrenal chromaffin granules and synaptic vesicles.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: solute carrier family 17 (vesicular nucleotide transporter), member 9; Solute carrier family 17 member 9; solute carrier family 17, member 9; Vesicular nucleotide transporter; vesicular nucleotide transporter SLC17A9; VNUT; Voltage-gated purine nucleotide uniporter SLC17A9
Gene Aliases: 1700019H03Rik; BC019537; C20orf59; POROK8; RGD1311940; SLC17A9; VNUT
UniProt ID: (Human) Q9BYT1, (Mouse) Q8VCL5, (Rat) P0DX21
Entrez Gene ID: (Human) 63910, (Mouse) 228993, (Rat) 362287
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support