Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
For reconstitution, we recommend adding 100 µL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiments, we strongly recommend performing another round of desalting. (Zeba Spin Desalting Columns, 7KMWCO, 0.5 mL, Product # 89882)
VMAT2 is involved in the ATP-dependent vesicular transport of biogenic amine neurotransmitters. pumps cytosolic monoamines including dopamine, norepinephrine, serotonin, and histamine into synaptic vesicles. It is a requisite for vesicular amine storage prior to secretion via exocytosis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MGC120477; MGC120478; MGC26538; monoamine neurotransmitter transporter; Monoamine transporter; solute carrier family 18 (vesicular monoamine transporter), member 2; solute carrier family 18 (vesicular monoamine), member 2; Solute carrier family 18 member 2; Synaptic vesicular amine transporter; VAT2; vesicle monoamine transporter type 2; vesicle monoamine/H+ antiporter; Vesicular amine transporter 2
Gene Aliases: SLC18A2; SVAT; SVMT; VAT2; VMAT2
UniProt ID: (Human) Q05940
Entrez Gene ID: (Human) 6571
Molecular Function:
secondary carrier transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support