Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 2
PA1-9553 detects SLC19A1 in human samples.
PA1-9553 has been successfully used in Western blot procedures. By Western blot, PA1-9553 detects a 65 kDa protein representing SLC19A1.
PA1-9553 immunogen is a purified recombinant SLC19A1 corresponding to the chromosome 21 gene (Human).
Transport of folate compounds into mammalian cells can occur via receptor-mediated or carrier-mediated mechanisms. A functional coordination between these 2 mechanisms has been proposed to be the method of folate uptake in certain cell types. Methotrexate (MTX) is an antifolate chemotherapeutic agent that is actively transported by the carrier-mediated uptake system. RFC1 plays a role in maintaining intracellular concentrations of folate.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cyclic dinucleotide:anion antiporter SLC19A1; Folate:anion antiporter SLC19A1; FOLT; hSLC19A1; IFC-1; Intestinal folate carrier 1; Placental folate transporter; Reduced folate carrier protein; Reduced folate transporter; Reduced folate transporter 1; RFC; RFT-1; solute carrier family 19 (folate transporter), member 1; Solute carrier family 19 member 1
Gene Aliases: CHMD; FLOT1; FOLT; IFC1; REFC; RFC1; SLC19A1
UniProt ID: (Human) P41440
Entrez Gene ID: (Human) 6573
Molecular Function: transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support