Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
The recommended shelf life for this product is 1 year from date of receipt.
Transporter for alanine, serine, cysteine, and threonine. Exhibits sodium dependence.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Alanine/serine/cysteine/threonine transporter 1; ASCT-1; glutamate/neutral amino acid transporter; Neutral amino acid transporter A; OTTHUMP00000159933; OTTHUMP00000235138; solute carrier family 1 (glutamate/neutral amino acid transporter), member 4; Solute carrier family 1 member 4
Gene Aliases: ASCT-1; ASCT1; AW045657; SATT; SLC1A4; SPATCCM
UniProt ID: (Human) P43007, (Mouse) O35874
Entrez Gene ID: (Human) 6509, (Mouse) 55963
Molecular Function: primary active transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support