Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MATLITSTTA ATAASGPLVD YLWMLILGFI IAFVLAFSVG ANDVANSFGT AVGSGVVTLK QACILASIFE TVGSVLLGAK VSETIRKGLI DVEMYNSTQG LLMAGSVSAM FGSAVWQLVA SFLKLPISGT HCIVGATIGF SLVAKGQEGV KWSELIKIVM SWFVSPLLSG IMSGILFFLV RAFILHKADP VPNGLRALPV FYACTVGINL FSIMYTGAPL LGFDKLPLWG TILISVGCAV FCALIVWFFV CPRMKRKIER EIKCSPSESP LMEKKNSLKE DHEETKLSVG DIENKHPVSE VGPATVPLQA VVEERTVSFK LGDLEEAPER ERLPSVDLKE ETSIDSTVNG AVQLPNGNLV QFSQAVSNQI NSSGHYQYHT VHKDSGLYKE LLHKLHLAKV GDCMGDSGDK PLRRNNSYTS YTMAICGMPL DSFRAKEGEQ KGEEMEKLTW PNADSKKRIR MDSYTSYCNA VSDLHSASEI DMSVKAEMGL GDRKGSNGSL EEWYDQDKPE VSLLFQFLQI LTACFGSFAH GGNDVSNAIG PLVALYLVYD TGDVSSKVAT PIWLLLYGGV GICVGLWVWG RRVIQTMGKD LTPITPSSGF SIELASALTV VIASNIGLPI STTHCKVGSV VSVGWLRSKK AVDWRLFRNI FMAWFVTVPI SGVISAAIMA IFRYVILRM
Retrovirus receptors allow infection of human and murine cells by various retroviruses. The receptors that have been identified at the molecular level include CD4 for human immunodeficiency virus, Rec1 for murine ecotropic virus, and GLVR1 for gibbon ape leukemia virus. These 3 proteins show no homology to one another at the DNA or protein level. GLVR1 is a sodium-dependent phosphate symporter.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DKFZp686J2397; FLJ41426; Gibbon ape leukemia virus receptor 1; GLVR-1; Leukemia virus receptor 1 homolog; Phosphate transporter 1; PiT-1; Sodium-dependent phosphate transporter 1; solute carrier family 20 (phosphate transporter), member 1; Solute carrier family 20 member 1
Gene Aliases: Glvr-1; GLVR1; PiT-1; PIT1; SLC20A1
UniProt ID: (Human) Q8WUM9
Entrez Gene ID: (Human) 6574
Molecular Function: transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support