Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: TGKLQKEGAL SRVSDESLSK VQEAESPVFK ELPGAKANDD STIPLTGAAG ETLGTSEGTS AGSHPRAAYG RALSMTHGSV KSPISNGTFG FDGHTRSDGH
This gene encodes a member of the inorganic phosphate transporter family. The encoded protein is a type 3 sodium-dependent phosphate symporter that plays an important role in phosphate homeostasis by mediating cellular phosphate uptake. The encoded protein also confers susceptibility to viral infection as a gamma-retroviral receptor. Mutations in this gene may play a role in familial idiopathic basal ganglia calcification. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Gibbon ape leukemia virus receptor 2; GLVR-2; hPit2; murine leukemia virus, amphotropic, receptor for; murine leukemia virus, amphotropic; receptor; Phosphate transporter 2; PiT-2; Sodium-dependent phosphate transporter 2; solute carrier family 20 (phosphate transporter), member 2; Solute carrier family 20 member 2
Gene Aliases: GLVR-2; GLVR2; IBGC1; IBGC3; MLVAR; PIT-2; PIT2; Ram-1; RAM1; SLC20A2
UniProt ID: (Human) Q08357
Entrez Gene ID: (Human) 6575
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support