Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: ESLRWLMAT QQFESAKRLI LHFTQKNRMN PEGDIKGVIP ELEKELSRRP KKVCIVKVVG TRNLWKNIVV LCVNSLTGYG IHHCFARSMM GHEVKVPLLE NFYA (84-186 aa encoded by BC128580)
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ion transporter protein; Solute carrier family 22 member 23; solute carrier family 22, member 23
Gene Aliases: 3110004L20Rik; C6orf85; Nritp; SLC22A23
UniProt ID: (Human) A1A5C7, (Mouse) Q3UHH2, (Rat) Q9QZG1
Entrez Gene ID: (Human) 63027, (Mouse) 73102, (Rat) 64559
Molecular Function:
secondary carrier transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support