Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: AAVPAHRCA LPGAPANFSH QDVWLEAHLP REPDGTLSSC LRFAYPQALP NTTLGEERQS RGELEDEPAT VPCSQGWEYD HSEFSSTIAT ESQWDLVCEQ (42-140 aa encoded by BC033805)
SLC22A7 is involved in the sodium-independent transport and excretion of organic anions, some of which are potentially toxic. The protein is an integral membrane protein and appears to be localized to the basolateral membrane of the kidney. The protein encoded by this gene is involved in the sodium-independent transport and excretion of organic anions, some of which are potentially toxic. The encoded protein is an integral membrane protein and appears to be localized to the basolateral membrane of the kidney. Alternatively spliced transcript variants encoding different isoforms have been described. The protein encoded by this gene is involved in the sodium-independent transport and excretion of organic anions, some of which are potentially toxic. The encoded protein is an integral membrane protein and appears to be localized to the basolateral membrane of the kidney. Alternatively spliced transcript variants encoding different isoforms have been described.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hOAT2; liver-specific transporter; MGC24091; MGC45202; mOAT2; NLT; Novel liver transporter; Organic anion transporter 2; rOAT2; solute carrier family 22 (organic anion transporter), member 7; Solute carrier family 22 member 7
Gene Aliases: NLT; OAT2; SLC22A7
UniProt ID: (Human) Q9Y694, (Rat) Q5RLM2, (Mouse) Q91WU2
Entrez Gene ID: (Human) 10864, (Rat) 89776, (Mouse) 108114
Molecular Function: secondary carrier transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support