Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Aliquoting is unnecessary for -20°C storage.
The mitochondrial tricarboxylate transporter (also called citrate transport protein, or CTP) is responsible for the movement of citrate across the mitochondrial inner membrane.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CIC; Citrate carrier; Citrate transport protein; citrate transporter) member 1; CTP; DiGeorge syndrome gene j; Mitochondrial citrate carrier; mitochondrial tricarboxylate carrier; solute carrier family 20 (mitochondrial citrate transporter), member 3; solute carrier family 25 (mitochondrial carrier; citrate transporter), member 1; Solute carrier family 25 member 1; Tricarboxylate carrier protein; Tricarboxylate transport protein, mitochondrial; txtp
Gene Aliases: 1300019P08Rik; 2610100G11Rik; AI194714; Cic; CTP; D2L2AD; Dgsj; SEA; SLC20A3; SLC25A1
UniProt ID: (Human) P53007, (Rat) P32089, (Mouse) Q8JZU2
Entrez Gene ID: (Human) 6576, (Rat) 29743, (Mouse) 13358
Molecular Function: transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support