Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MAAEARVSR WYFGGLASCG AACCTHPLDL LKVHLQTQQE VKLRMTGMAL RVVRTDGILA LYSGLSASLC RQMTYSLTRF AIYETVRDRV AKGSQGPLPF HEKVLLGSVS GLAGGFVGTP ADLVNVRMQN DVKLPQGQRR NYAHALDGLY RVAREEGLRR LFSGATMASS RGALVTVGQL SCYDQAKQLV LSTGYLSDNI FTHFVASFIA AAGDEPPPQG GCATFLCQPL DVLKTRLMNS KGEYQGVFHC AVETAKLGPL AFYKGLVPAG IRLIPHTVLT FVFLEQLRKN FGIKVPS (1-296 aa encoded by BC015797)
The mitochondrial dicarboxylate carrier protein (mDIC) belongs to superfamily of mitochondrial transporters. It has been demonstrated that the expression of mDIC leads to hyperpolarization of the mitochondria, as well as succinate uptake by the mitochondria. It is highly expressed in adipose tissue, where its expression is downregulated by insulin and upregulated by free fatty acids. mDIC is also thought to play a role in glyceroneogenesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DIC; dicarboxylate ion carrier; dicarboxylate transporter), member 10; Mitochondrial dicarboxylate carrier; solute carrier family 25 (mitochondrial carrier; adenine nucleotide translocator), member 10; solute carrier family 25 (mitochondrial carrier; dicarboxylate transporter), member 10; Solute carrier family 25 member 10
Gene Aliases: DIC; SLC25A10
UniProt ID: (Human) Q9UBX3, (Mouse) Q9QZD8, (Rat) O89035
Entrez Gene ID: (Human) 1468, (Mouse) 27376, (Rat) 170943
Molecular Function:
transporter
secondary carrier transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support