Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide sequence: GLKPAGSEPT PKSRIADLPP ANPDHIGGYR LATATFAGIE NKFGLYLPKF
Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%
This gene encodes a calcium-binding mitochondrial carrier protein. The encoded protein localizes to the mitochondria and is involved in the exchange of aspartate for glutamate across the inner mitochondrial membrane. Polymorphisms in the gene encoding this protein may be associated with risk of autism.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Araceli hiperlarga; Aralar; calcium binding mitochondrial carrier superfamily member Aralar1; Electrogenic aspartate/glutamate antiporter SLC25A12, mitochondrial; Mitochondrial aspartate glutamate carrier 1; solute carrier family 25 (aspartate/glutamate carrier), member 12; solute carrier family 25 (mitochondrial carrier, Aralar), member 12; Solute carrier family 25 member 12
Gene Aliases: AGC1; ARALAR; ARALAR1; SLC25A12
UniProt ID: (Human) O75746
Entrez Gene ID: (Human) 8604
Molecular Function: transporter secondary carrier transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support