Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Immunogen sequence: MKSNPAIQA AIDLTAGAAG GTACVLTGQP FDTMKVKMQT FPDLYRGLTD CCLKTYSQVG FRGFYKGTSP ALIANIAENS VLFMCYGFCQ QVVRKVAGLD KQAKLSDLQN AAAGSFASAF AALVLCPTEL VKCRLQTMYE METSGKIAKS QNTVWSVIKS ILRKDGPLGF YHGLSSTLLR EVPGYFFFFG GYELSRSFFA SGRSKDELGP VPLMLSGGVG GICLWLAVYP VDCIKSRIQV LSMSGKQAGF IRTFLNVVKN EGITALYSGL KPTMIRAFPA NGALFLAYEY SRKLMMNQLE AY (1-301 aa encoded by BC002702)
The SLC25A15 gene encodes the mitochondrial ornithine transporter, which transports ornithine across the inner mitochondrial membrane from the cytosol to the mitochondrial matrix. This is a vital step in the urea cycle, which serves to eliminate toxic ammonium ions from the breakdown of nitrogen.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Mitochondrial ornithine transporter 1; ornithine transporter 1; ornithine transporter) member 15; ornithine transporter, mitochondrial 1; solute carrier family 25 (mitochondrial carrier; ornithine transporter) member 15; Solute carrier family 25 member 15
Gene Aliases: Ab1-114; D13S327; D630044L02Rik; HHH; ORC1; ORNT1; SLC25A15; SP1855
UniProt ID: (Human) Q9Y619, (Rat) A0A0G2K309, (Mouse) Q9WVD5
Entrez Gene ID: (Human) 10166, (Rat) 306574, (Mouse) 18408
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support