Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Immunogen sequence: MRGSPGDAE RRQRWGRLFE ELDSNKDGRV DVHELRQGLA RLGGGNPDPG AQQGISSEGD ADPDGGLDLE EFSRYLQERE QRLLLMFHSL DRNQDGHIDV SEIQQSFRAL GISISLEQAE KILHSMDRDG TMTIDWQEWR DHFLLHSLEN VEDVLYFWKH STLSSAGFSA WIKDSTAEQN RSKTTVLARR (1-189 aa encoded by BC001656)
Calcium-dependent mitochondrial solute carrier. Mitochondrial solute carriers shuttle metabolites, nucleotides, and cofactors through the mitochondrial inner membrane (PubMed:15123600). May act as a ATP-Mg/Pi exchanger that mediates the transport of Mg-ATP in exchange for phosphate, catalyzing the net uptake or efflux of adenine nucleotides into or from the mitochondria (PubMed:15123600). Acts as a regulator of mitochondrial calcium uptake via interaction with MCU and MICU1 (PubMed:24430870). [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: calcium-binding mitochondrial carrier protein SCaMC-3; Mitochondrial adenyl nucleotide antiporter SLC25A23; Mitochondrial ATP-Mg/Pi carrier protein 2; mitochondrial Ca(2+)-dependent solute carrier protein 2; mitochondrial Ca2+-dependent solute carrier protein 2; phosphate carrier), member 23; SCaMC-3; short calcium-binding mitochondrial carrier 3; Short calcium-binding mitochondrial carrier protein 3; sma; small calcium-binding mitochondrial carrier protein 3; solute carrier family 25 (mitochondrial carrier; phosphate carrier), member 23; Solute carrier family 25 member 23
Gene Aliases: 2310067G05Rik; APC2; MCSC2; SCaMC-3; SCAMC3; SLC25A23
UniProt ID: (Human) Q9BV35, (Mouse) Q6GQS1
Entrez Gene ID: (Human) 79085, (Rat) 301113, (Mouse) 66972
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support