Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: P5906365 Don't miss out: buy 3, only pay for 2 Extended to June 27: take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Positive Samples: HeLa, 293T, Mouse pancreas, Mouse testis, Mouse brain, Mouse kidney, Rat testis, Rat brain
Immunogen sequence: MRGSPGDAER RQRWGRLFEE LDSNKDGRVD VHELRQGLAR LGGGNPDPGA QQGISSEGDA DPDGGLDLEE FSRYLQEREQ RLLLMFHSLD RNQDGHIDVS EIQQSFRALG ISISLEQAEK ILHSMDRDGT MTIDWQEWRD HFLLHSLENV EDVLYFWKHS
Calcium-dependent mitochondrial solute carrier. Mitochondrial solute carriers shuttle metabolites, nucleotides, and cofactors through the mitochondrial inner membrane (PubMed:15123600). May act as a ATP-Mg/Pi exchanger that mediates the transport of Mg-ATP in exchange for phosphate, catalyzing the net uptake or efflux of adenine nucleotides into or from the mitochondria (PubMed:15123600). Acts as a regulator of mitochondrial calcium uptake via interaction with MCU and MICU1 (PubMed:24430870). [UniProt]
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: calcium-binding mitochondrial carrier protein SCaMC-3; Mitochondrial adenyl nucleotide antiporter SLC25A23; Mitochondrial ATP-Mg/Pi carrier protein 2; mitochondrial Ca(2+)-dependent solute carrier protein 2; mitochondrial Ca2+-dependent solute carrier protein 2; phosphate carrier), member 23; SCaMC-3; short calcium-binding mitochondrial carrier 3; Short calcium-binding mitochondrial carrier protein 3; sma; small calcium-binding mitochondrial carrier protein 3; solute carrier family 25 (mitochondrial carrier; phosphate carrier), member 23; Solute carrier family 25 member 23
Gene Aliases: 2310067G05Rik; APC2; MCSC2; SCaMC-3; SCAMC3; SLC25A23
UniProt ID: (Human) Q9BV35, (Mouse) Q6GQS1
Entrez Gene ID: (Human) 79085, (Rat) 301113, (Mouse) 66972
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support