Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MATGGQQKE NTLLHLFAGG CGGTVGAIFT CPLEVIKTRL QSSRLALRTV YYPQVHLGTI SGAGMVRPTS VTPGLFQVLK SILEKEGPKS LFRGLGPNLV GVAPSRAVYF ACYSKAKEQF NGIFVPNSNI VHIFSAGSAA FITNSLMNPI WMVKTRMQLE QKVRGSKQMN TLQCARYVYQ TEGIRGFYRG LTASYAGISE TIICFAIYES LKKYLKEAPL ASSANGTEKN STSFFGLMAA AALSKGCASC IAYPHEVIRT RLREEGTKYK SFVQTARLVF REEGYLAFYR GLFAQLIRQI PNTAIVLSTY ELIVYLLEDR TQ (1-321 aa encoded by BC073135)
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BMSC-MCP; Bone marrow stromal cell mitochondrial carrier protein; huBMSC-MCP; mitochondrial carrier protein; novel mitochondrial carrier protein; PNC1 protein; Protein PNC1; solute carrier family 25 (pyrimidine nucleotide carrier), member 33; Solute carrier family 25 member 33; solute carrier family 25, member 33
Gene Aliases: BMSC-MCP; PNC1; SLC25A33
UniProt ID: (Human) Q9BSK2
Entrez Gene ID: (Human) 84275
Molecular Function: mitochondrial carrier protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support