Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
FIGURE: 1 / 1
This antibody detects human FATP1 in cell transfectants but not non-transfected parent cells in flow cytometry.
Reconstitute at 0.5 mg/mL in sterile PBS.
SLC27A1 is a protein coding gene. Mediates the ATP-dependent import of long-chain fatty acids (LCFA) into the cell by mediating their translocation at the plasma membrane. Has also an acyl-CoA ligase activity for long-chain and very-long-chain fatty acids. May act directly as a bona fide transporter, or alternatively, in a cytoplasmic or membrane-associated multimeric protein complex to trap and draw fatty acids towards accumulation. Plays a pivotal role in regulating available LCFA substrates from exogenous sources in tissues undergoing high levels of beta-oxidation or triglyceride synthesis. May be involved in regulation of cholesterol metabolism.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ACSVL5FATP-1; Arachidonate--CoA ligase; EC 6.2.1; FATP-1; Fatty acid transport protein 1; FLJ00336; Long-chain fatty acid transport protein 1; Long-chain-fatty-acid--CoA ligase; MGC71751; solute carrier family 27 (fatty acid transporter), member 1; Solute carrier family 27 member 1; Very long-chain acyl-CoA synthetase
Gene Aliases: ACSVL5; FATP; FATP-1; FATP1; SLC27A1
UniProt ID: (Human) Q6PCB7
Entrez Gene ID: (Human) 376497
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support