Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: GMAALVLRPP HALDLMQLYT HVSENLPPYA RPRFLRLQES LATTETFKQQ KVRMANEGFD PSTLSDPLYV LDQAVGAYLP LTTARYSALL AGNLRI
SLC27A3 has acyl-CoA ligase activity for long-chain and very-long-chain fatty acids.SLC27A3 does not exhibit fatty acid transport activity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Arachidonate--CoA ligase; FATP-3; fatty acid transport protein 3; Long-chain fatty acid transport protein 3; Long-chain-fatty-acid--CoA ligase; solute carrier family 27 (fatty acid transporter), member 3; Solute carrier family 27 member 3; Very long-chain acyl-CoA synthetase homolog 3; VLCS-3
Gene Aliases: ACSVL3; FATP3; PSEC0067; SLC27A3; UNQ367/PRO703; VLCS-3
UniProt ID: (Human) Q5K4L6
Entrez Gene ID: (Human) 11000
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support