Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This antibody detects endogenous protein at a molecular weight of 71 kDa.
Purity is >95% by SDS-PAGE.
Nucleosides play a role in signaling in several physiologic systems, and synthetic analogs of natural nucleosides are often used to treat neoplastic and viral diseases. Plasma membrane transport of nucleosides is mediated by equilibrative and concentrative nucleoside transporters, which may have specificity for purines or pyrimidines. The deduced human 650 amino acid concentrative nucleoside transporter 1 (CNT1) protein is 83% identical to the rat protein and is expressed in the intestine, kidney and liver. CNT1, also designated solute carrier family 28 (sodium-coupled nucleoside transporter), member 1 (SLC28A1), expedites sodium-dependent fluxes of uridine, azidodeoxythymidine (AZT) and adenosine, but not of guanosine or deoxyadenosine, which undergo net renal secretion. CNT1 activity may serve as a putative mechanism for renal reabsorption of physiologic nucleosides and synthetic nucleoside drugs.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CNT 1; Concentrative nucleoside transporter 1; Na(+)/nucleoside cotransporter 1; Sodium-coupled nucleoside transporter 1; Sodium/nucleoside cotransporter 1; solute carrier family 28 (concentrative nucleoside transporter), member 1; solute carrier family 28 (sodium-coupled nucleoside transporter), member 1; Solute carrier family 28 member 1; solute carrier family 28, member 1
Gene Aliases: CNT1; HCNT1; SLC28A1
UniProt ID: (Human) O00337, (Rat) Q62674, (Mouse) E9PXX9
Entrez Gene ID: (Human) 9154, (Rat) 116642, (Mouse) 434203
Molecular Function:
transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support