Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Reconstitute in 100 µL of sterile water. Centrifuge to remove any insoluble material.
The peptide is homologous in rat and mouse.
Specificity of this antibody: SLC30A5.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hZTL1; Proton-coupled zinc antiporter SLC30A5; solute carrier family 30 (zinc transporter), member 5; Solute carrier family 30 member 5; Zinc transporter 5; zinc transporter like 1; zinc transporter ZTL1; ZnT-5; ZnT-like transporter 1
Gene Aliases: 1810010K08Rik; AF233321; SLC30A5; UNQ863/PRO1879; ZnT-5; ZNT5; ZNTL1; ZTL1
UniProt ID: (Human) Q8TAD4, (Mouse) Q8R4H9
Entrez Gene ID: (Human) 64924, (Mouse) 69048, (Rat) 294698
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support