Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MAAPRDNVT LLFKLYCLAV MTLMAAVYTI ALRYTRTSDK ELYFSTTAVC ITEVIKLLLS VGILAKETGS LGRFKASLRE NVLGSPKELL KLSVPSLVYA VQNNMAFLAL SNLDAAVYQV TYQLKIPCTA (1-129 aa encoded by BC017807)
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CMP-SA-Tr; CMP-Sia-Tr; CMP-sialic acid transporter; mutated CMP-sialic acid transporter A1; solute carrier family 35 (CMP-sialic acid transporter), member 1; solute carrier family 35 (CMP-sialic acid transporter), member A1; solute carrier family 35 (UDP-galactose transporter), member 1; Solute carrier family 35 member A1
Gene Aliases: AA408150; AI314851; CDG2F; CMPST; CST; hCST; SLC35A1
UniProt ID: (Human) P78382, (Mouse) Q61420
Entrez Gene ID: (Human) 10559, (Rat) 313139, (Mouse) 24060
Molecular Function: transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support