Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: VLTVHCMVIL LNCAQHLSQR LQKTFVNYGE ATMYGLETCP NTWLRAHAVW GRWNLALSPR LECSGKISAH CNPHLQGSSN SPAQASRVAG IYRYTVSFLL VITQLGF
The function remains unknown.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Proton-coupled amino acid transporter 3; Proton/amino acid transporter 3; solute carrier family 36 (proton/amino acid symporter), member 3; Solute carrier family 36 member 3; solute carrier family 36, member 3; tramdorin 2; Tramdorin-2
Gene Aliases: PAT3; SLC36A3; TRAMD2; tramdorin2
UniProt ID: (Human) Q495N2, (Mouse) Q811P0
Entrez Gene ID: (Human) 285641, (Mouse) 215332
Molecular Function: amino acid transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support