Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: DVRCSSTLV THSKGYENGT NRLRLQKQIL KSEKNKPLDP EMQCLLLSDG KGSIHPNHVV ILPGDGGSGT AAISFTGALK IPGVIEFSLC L (247-336 aa encoded by BC152997)
SLC37A1, a member of the sugar-phosphate transport family, transports glycerol-3-phosphate (G3P) between cellular compartments for its utilization in several compartment-specific biochemical pathways.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ22340; G-3-P permease; G-3-P transporter; Glucose-6-phosphate exchanger SLC37A1; Glycerol-3-phosphate permease; glycerol-3-phosphate transporter; solute carrier family 37 (glucose-6-phosphate transporter), member 1; solute carrier family 37 (glycerol-3-phosphate transporter), member 1; Solute carrier family 37 member 1
Gene Aliases: G3PP; SLC37A1
UniProt ID: (Human) P57057, (Mouse) Q8R070
Entrez Gene ID: (Human) 54020, (Mouse) 224674
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support