Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MAWPNVFQRG SLLSQFSHHH VVVFLLTFFS YSLLHASRKT FSNVKVSISE QWTPSAFNTS VELPVEIWSS NHLFPSAEKA TLFLGTLDTI FLFSYAVGLF ISGIVGDRLN LRWVLSFGMC SSALVVFVFG ALTEWLRFYN KWLYCCLWIV NGLLQSTGWP CVVAVMGNWF GKAGRGVVFG LWSACASVGN ILGACLASSV LQYGYEYAFL VTASVQFAGG IVIFFGLLVS PEEIGLSGIE AEENFEEDSH RPLINGGENE DEYEPNYSIQ DDSSVAQVKA ISFYQACCLP GVIPYSLAYA CLKLVNYSFF FWLPFYLSNN FGWKEAEADK LSIWYDVGGI IGGTLQGFIS DVLQKRAPVL ALSLLLAVGS LIGYSRSPND KSINALLMTV TGFFIGGPSN MISSAISADL GRQELIQRSS EALATVTGIV DGSGSIGAAV GQYLVSLIRD KLGWMWVFYF FILMTSCTIV FISPLIVREI FSLVLRRQAH ILRE
SLC37A3 (Solute Carrier Family 37 Member 3) is a Protein Coding gene. Diseases associated with SLC37A3 include Glycerol Kinase Deficiency and Deafness, Autosomal Recessive 8. Gene Ontology (GO) annotations related to this gene include transporter activity and transmembrane transporter activity. An important paralog of this gene is SLC37A1. [GeneCards]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: solute carrier family 37 (glycerol-3-phosphate transporter), member 3; Solute carrier family 37 member 3; solute carrier family 37, member 3; Sugar phosphate exchanger 3
Gene Aliases: SLC37A3; SPX3
UniProt ID: (Human) Q8NCC5
Entrez Gene ID: (Human) 84255
Molecular Function:
transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support