Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
Antibody detects endogenous levels of total SLC38A3.
As a sodium-dependent amino acid/proton antiporter, SLC38A3 mediates electrogenic cotransport of glutamine and sodium ions in exchange for protons. It also recognizes histidine, asparagine and alanine. It may mediate amino acid transport in either direction under physiological conditions and play a role in nitrogen metabolism and synaptic transmission.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: mNAT; N system amino acids transporter NAT-1; N-system amino acid transporter 1; Na(+)-coupled neutral amino acid transporter 3; SLC38A3; slc38a3 {ECO:0000312|RGD:628620}; Sodium-coupled neutral amino acid transporter 3; Solute carrier family 38 member 3; solute carrier family 38, member 3; System N amino acid transporter 1; system N1 amino acid transporter; system N1 Na+ and H+-coupled glutamine transporter
Gene Aliases: 0610012J02Rik; D9Ucla2; G17; mNAT; NAT1; Slc38-3; SLC38A3; SN1; SNAT3
UniProt ID: (Human) Q99624, (Rat) Q9JHZ9, (Mouse) Q9DCP2
Entrez Gene ID: (Human) 10991, (Rat) 252919, (Mouse) 76257
Molecular Function: amino acid transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support