Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Immunogen sequence: MDPMELRNV NIEPDDESSS GESAPDSYIG IGNSEKAAMS SQFANEDTES QKFLTNGFLG KKKLADYADE HHPGTTSF (1-77 aa encoded by BC101827)
SLC38A4 is found predominantly in liver and transports both cationic and neutral amino acids. The transport of cationic amino acids by SLC38A4 is Na(+) and pH independent, while the transport of neutral amino acids is Na(+) and pH dependent.The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes, or E1s, ubiquitin-conjugating enzymes, or E2s, and ubiquitin-protein ligases, or E3s. This gene encodes a member of the E2 ubiquitin-conjugating enzyme family. Four alternatively spliced transcript variants encoding the same protein have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Amino acid transporter A3; amino acid transporter system A3; FLJ10191; MGC126876; N amino acid transporter 3; Na(+)-coupled neutral amino acid transporter 4; Sodium-coupled neutral amino acid transporter 4; Solute carrier family 38 member 4; solute carrier family 38, member 4; System A amino acid transporter 3; System N amino acid transporter 3
Gene Aliases: ATA3; NAT3; PAAT; SLC38A4; SNAT4
UniProt ID: (Human) Q969I6
Entrez Gene ID: (Human) 55089
Molecular Function:
amino acid transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support