Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: QQDKIIAVMA KEPEGASGPW YTDRK
Antigen sequence identity: Mouse 88%; Rat 92%
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: amino acid transporter; Sodium-coupled neutral amino acid transporter 7; Solute carrier family 38 member 7; solute carrier family 38, member 7
Gene Aliases: SLC38A7; SNAT7
UniProt ID: (Human) Q9NVC3
Entrez Gene ID: (Human) 55238
Molecular Function: amino acid transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support