Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Immunogen sequence: MANMNSDSRH LGTSEVDHER DPGPMNIQFE PSDLRSKRPF CIEPTNIVNV NHVIQRVSDH ASAMNKRIHY YSRLTTPADK ALIPPDHVVP APEECYVYSP LGSAYKLQSY TEGYGKNTSL VTIFMIWNTM M
SLC38A9 is a protein coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Neutral amino acid transporter 9; putative sodium-coupled neutral amino acid transporter 9; sodium-coupled neutral amino acid transporter 9; Solute carrier family 38 member 9; solute carrier family 38, member 9; Up-regulated in lung cancer 11
Gene Aliases: 4833412L08Rik; 6720411P22Rik; 9130023D20Rik; 9430067K09Rik; A730092C09; SLC38A9; URLC11
UniProt ID: (Human) Q8NBW4, (Mouse) Q8BGD6
Entrez Gene ID: (Human) 153129, (Mouse) 268706
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support